PloS one
Identification of Anti-EGFR and Anti-ErbB3 Dual Variable Domains Immunoglobulin (DVD-Ig) Proteins with Unique Activities.
PMID 25997020
Abstract
Epidermal growth factor receptor (EGFR) and receptor tyrosine-protein kinase 3 (ErbB3) are two well-established targets in cancer therapy. There is significant crosstalk among these two receptors and others. To block signaling from both EGFR and ErbB3, we generated anti-EGFR and anti-ErbB3 DVD-Ig proteins. Two DVD-Ig proteins maintained the functions of the combination of the two parental antibodies. The DVD-Ig proteins inhibit cell signaling and proliferation in A431 and FaDu cells while half DVD-Ig proteins lost proliferation inhibition function. Interestingly, in the presence of β-Heregulin (HRG), the DVD-Ig proteins show synergies with respect to inhibiting cell proliferation. The DVD-Ig proteins downregulate EGFR protein expression in the presence of HRG, which may be due to receptor internalization. Furthermore, the DVD-Ig proteins remarkably disrupt β-Heregulin binding to FaDu cells.