EMAIL THIS PAGE TO A FRIEND
Molecular autism
NTRK2 expression levels are reduced in laser captured pyramidal neurons from the anterior cingulate cortex in males with autism spectrum disorder.
PMID 26000162
Abstract
The anterior cingulate cortex (ACC) is a brain area involved in modulating behavior associated with social interaction, disruption of which is a core feature of autism spectrum disorder (ASD). Functional brain imaging studies demonstrate abnormalities of the ACC in ASD as compared to typically developing control patients. However, little is known regarding the cellular basis of these functional deficits in ASD. Pyramidal neurons in the ACC are excitatory glutamatergic neurons and key cellular mediators of the neural output of the ACC. This study was designed to investigate the potential role of ACC pyramidal neurons in ASD brain pathology. Postmortem ACC tissue from carefully matched ASD and typically developing control donors was obtained from two national brain collections. Pyramidal neurons and surrounding astrocytes were separately collected from layer III of the ACC by laser capture microdissection. Isolated RNA was subjected to reverse transcription and endpoint PCR to determine gene expression levels for 16 synaptic genes relevant to glutamatergic neurotransmission. Cells were also collected from the prefrontal cortex (Brodmann area 10) to examine those genes demonstrating differences in expression in the ACC comparing typically developing and ASD donors. The level of NTRK2 expression was robustly and significantly lower in pyramidal neurons from ASD donors as compared to typically developing donors. Levels of expression of GRIN1, GRM8, SLC1A1, and GRIP1 were modestly lower in pyramidal neurons from ASD donors, but statistical significance for these latter genes did not survive correction for multiple comparisons. No significant expression differences of any genes were found in astrocytes laser captured from the same neocortical area. In addition, expression levels of NTRK2 and other synaptic genes were normal in pyramidal neurons laser captured from the prefrontal cortex. These studies demonstrate a unique pathology of neocortical pyramidal neurons of the ACC in ASD. NTRK2 encodes the tropomyosin receptor kinase B (TrkB), transmission through which neurotrophic factors modify differentiation, plasticity, and synaptic transmission. Reduced pyramidal neuron NTRK2 expression in the ACC could thereby contribute to abnormal neuronal activity and disrupt social behavior mediated by this brain region.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A1028
8-Anilino-1-naphthalenesulfonic acid
C16H13NO3S