EMAIL THIS PAGE TO A FRIEND
Testing
Mitochondrial Fragmentation Due to Inhibition of Fusion Increases Cyclin B through Mitochondrial Superoxide Radicals.
PMID 26000631
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
06-570
Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker