EMAIL THIS PAGE TO A FRIEND
Journal of colloid and interface science
Jellyfish mesoglea as a matrix for the synthesis of extremely high content silver dendrites.
PMID 26001133
Abstract
Using macroporous jellyfish mesoglea which consists of nano-sized layers and fibers as a matrix, metal nanoparticles with different morphologies might be formed in it. Hierarchical silver dendrites are prepared by in situ reduction of silver nitrate (AgNO3) in the jellyfish mesoglea. The formation of silver dendrites is affected by the concentration of AgNO3. The biogel-metal nanoparticles hybrid materials with silver dendrites show an extremely high solid content of silver dendrites and good catalytic activity as well as excellent recyclability for the reduction of 4-nitrophenol (4-NP) to 4-aminophenol (4-AP).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
241326
4-Nitrophenol, ReagentPlus®, ≥99%
C6H5NO3
N7660
4-Nitrophenol solution, 10 mM
C6H5NO3