EMAIL THIS PAGE TO A FRIEND
European radiology
Differentiating malignant vertebral tumours from non-malignancies with CT spectral imaging: a preliminary study.
PMID 26002126
Abstract
To investigate the value of dual-energy spectral computed tomography (DESCT) for differentiating malignant vertebral tumours from non-malignancies during venous phase. This study was institutional review board-approved, and written informed consent was obtained from all patients. Thirty-seven patients were examined by DESCT during venous phase. Twenty patients had malignant vertebral tumours, 17 had non-malignant vertebral tumours. The iodine/water densities for the lesion, the lesion-to-muscle ratio, and lesion-to-artery ratio for iodine density measurements were calculated and compared between the two groups with the two-tailed Student t test. A p-value < 0.05 was considered statistically significant. Sensitivity and specificity were compared between the qualitative and quantitative studies. The iodine density, lesion-to-muscle ratio, and lesion-to-artery ratio of the iodine density measurement for malignant vertebral tumours were significantly different from the respective values for non-malignancies (all p < 0.05). Using 0.52 as the threshold value for the lesion-to-artery iodine density ratio, one could obtain sensitivity of 85% and specificity of 100% for differentiating malignant vertebral tumours from non-malignancies, significantly higher than the qualitative diagnosis. DESCT imaging enables analysis of a number of additional quantitative CT parameters to improve the accuracy for differentiating malignant vertebral tumours from non-malignancies during venous phase. • Dual-energy CT provides a new quantitative method. • CT spectral imaging improves the accuracy of differential diagnosis. • Lesion-to-artery iodine density ratio for malignant vertebral tumours was higher than for non-malignancies.