EMAIL THIS PAGE TO A FRIEND
Neurochemistry international
Serotonin abnormalities in Engrailed-2 knockout mice: New insight relevant for a model of Autism Spectrum Disorder.
PMID 26002543
Abstract
Autism spectrum disorder (ASD) is a congenital neurodevelopmental behavioral disorder that appears in early childhood. Recent human genetic studies identified the homeobox transcription factor, Engrailed 2 (EN2), as a possible ASD susceptibility gene. En2 knockout mice (En2-/-) display subtle cerebellar neuropathological changes and reduced levels of tyrosine hydroxylase, noradrenaline and serotonin in the hippocampus and cerebral cortex similar to those ones which have been observed in the ASD brain. Furthermore other similarities link En2 knockout mice to ASD patients. Several lines of evidence suggest that serotonin may play an important role in the pathophysiology of the disease. In the present study we measured, by using an HPLC, the 5-HT levels in different brain areas and at different ages in En2-/- mice. In the frontal and occipital cortex, the content of 5HT was reduced in En2-/- 1 and 3 months old mice; in 6 month old mice, the difference was still present, but it was not statistically significant. The 5-HT content of cerebellar cortex was significantly reduced at 1 month old but significantly high when the KO mice reached 3 months of age. The increase was present even at 6 months of age. A similar trend was highlighted by SERT immunolabeling in En2-/- mice compared to control in the same areas and age analyzed. Our findings, in agreement with the current knowledge on the 5-HT system alterations in ASD, confirm the early neurotransmitter deficit with a late compensatory recovery in En2 KO-mice further suggesting that this experimental animal may be considered a good predictive model for the human disease.