EMAIL THIS PAGE TO A FRIEND
Human reproduction (Oxford, England)
NEDD8-mediated neddylation is required for human endometrial stromal proliferation and decidualization.
PMID 26003431
Abstract
Does NEDD8-mediated neddylation regulate human endometrial stromal proliferation and decidualization? Neddylation inhibition by a selective NEDD8-activating enzyme inhibitor, MLN4924, significantly impairs human endometrial stromal cell (HESC) proliferation and decidualization and facilitates cell senescence, via p21 accumulation. Neddylation regulates cell proliferation and tissue remodeling during embryogenesis and tumorigenesis, while human endometrial stroma undergoes sequential proliferation, differentiation, as well as dynamic tissue remodeling during each menstrual cycle. We first analyzed the expression of NEDD8 in human endometrial tissues from 50 subjects, and then explored the consequence of neddylation inhibition by MLN4924 on HESCs proliferation, decidualization and cellular senescence. We collected 50 dated human endometrial tissues from early proliferative stage to late secretory phase of the menstrual cycle and analyzed the NEDD8 expression and cellular location in human endometrium by employing quantitative real-time PCR (qRT-PCR) and immunohistochemistry staining. Similar approaches were also used to explore the mRNA and protein expression of NEDD8 in an immortalized human endometrial stromal cell line (HESC) during proliferation and decidualization (N = 6). An MTS assay was performed to evaluate the effects of neddylation inhibition by MLN4924 on HESC proliferation. Flow cytometry and BrdU incorporation assay were conducted to determine the HESC cell cycle progression in response to MLN4924 exposure during proliferation. We also analyzed F-actin distribution by phalloidin staining and decidual marker gene expression by qRT-PCR to accesses the consequence of neddylation inhibition on HESC decidualization. Immunoblotting analysis of cullin1 and p21, and SA-β-Galactosidase staining were performed to reveal the potential molecular basis for the impaired HESC proliferation, decidualization and cellular senescence. The siRNA technique was applied to knockdown p21 expression to test whether a clearance of p21 accumulation would correct the HESC defects from neddylation inhibition. We demonstrated that NEDD8 is ubiquitously expressed in human endometrium including luminal epithelium, glandular epithelium and the stromal cells during the menstrual cycle, as well as in the HESCs during proliferation and differentiation in culture. Employing multiple molecular, cellular and pharmacological approaches, we further observed that neddylation inhibition by MLN4924 significantly attenuates HESC proliferation (P-value < 0.05), impairs decidual transformation (P-value < 0.05), and facilitates cellular senescence. These abnormal HESC activities upon MLN4924 exposure were accompanied with reduced cullin1 neddylation and an aberrant accumulation of p21. While a clearance of p21 accumulation by siRNA knockdown could partially restore HESC proliferation and cellular viability, it failed to correct the decidualization defects. Since NEDD8 was also intensely expressed in the endometrial epithelium, it is interesting to further study its potential role in stroma-epithelial interactions through isolating and culturing epithelial cells. p21 siRNA knockdown experiments revealed that there are differential molecular machineries, other than p21, that are subject to neddylation regulation during HESC proliferation compared with differentiation. This alternative mechanism warrants further investigation in future. Our findings add novel evidence showing, for what we believe the first time, that NEDD8-mediated neddylation is required for normal human endometrial functions, which raises the possibility of approaching the neddylation system for diagnosis and treatment of infertility in women. This work was supported in parts by the National Basic Research Program of China (2011CB944400 to H.W.) and the National Natural Science Foundation (81130009, 81330017 to H.W., 81170575 to S.Q. and 31471106 to S.Z.). The author declares that there is no conflict of interest.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A9501
Adenosine 3′,5′-cyclic monophosphate, ≥98.5% (HPLC), powder
C10H12N5O6P
A3262
Adenosine 3′,5′-cyclic monophosphate tris salt, ≥97% (HPLC), powder
C10H12N5O6P · C4H11NO3
P9346
Liquified Phenol, ≥89.0%
C6H6O
M1629
Medroxyprogesterone 17-acetate, ≥97% (HPLC)
C24H34O4
P1037
Phenol, for molecular biology
C6H6O
P5566
Phenol, BioXtra, ≥99.5% (GC)
C6H6O
328111
Phenol, unstabilized, purified by redistillation, ≥99%
C6H6O
242322
Phenol, contains hypophosphorous as stabilizer, loose crystals, ACS reagent, ≥99.0%
C6H6O
185450
Phenol, unstabilized, ReagentPlus®, ≥99%
C6H6O
77607
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
C6H6O
33517
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
C6H6O
16016
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached)
C6H6O
16017
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
C6H6O
16018
Phenol, ≥96.0% (calc. on dry substance, T)
C6H6O
W322318
Phenol, ≥99%
C6H6O
W322340
Phenol, natural, 97%, FG
C6H6O
P4557
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, for molecular biology
C6H6O
P4682
Phenol solution, Saturated with 0.1 M citrate buffer, pH 4.3 ± 0.2, BioReagent, for molecular biology
C6H6O
484431
Propylene glycol monomethyl ether acetate, ReagentPlus®, ≥99.5%
C6H12O3
537543
Propylene glycol monomethyl ether acetate, 99%
C6H12O3
209643
Selenium, pellets, <5 mm, ≥99.99% trace metals basis
Se
204307
Selenium, pellets, <5 mm particle size, ≥99.999% trace metals basis
Se
229865
Selenium, powder, −100 mesh, 99.99% trace metals basis
Se
209651
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Se
HT-SE101
Selenium, pellets, < 5mm, ≥99.999%
Se
GF66395212
Selenium, foil, 25x25mm, thickness 3mm, 99.95%
Se
S6297
Sodium bicarbonate, BioXtra, 99.5-100.5%
CHNaO3
S4019
Sodium bicarbonate, Hybri-Max, powder, suitable for hybridoma, ≥99.5%
CHNaO3
792500
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
C3H3NaO3
80443
Sodium pyruvate, PharmaGrade, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production.
C3H3NaO3