EMAIL THIS PAGE TO A FRIEND
Talanta
Simultaneous speciation of inorganic arsenic, selenium and tellurium in environmental water samples by dispersive liquid liquid microextraction combined with electrothermal vaporization inductively coupled plasma mass spectrometry.
PMID 26003714
Abstract
A new method based on dispersive liquid liquid microextraction (DLLME) combined with electrothermal vaporization inductively coupled plasma mass spectrometry (ETV-ICP-MS) was developed for the simultaneous speciation of inorganic arsenic (As), selenium (Se) and tellurium (Te) with sodium diethyldithiocarbamate (DDTC) as both chelating reagent and chemical modifier. As(III), Se(IV) and Te(IV) were transformed into DDTC-chelates at pH 7 and extracted into the fine droplets formed by injecting the binary solution of bromobenzene (extraction solvent) and methanol (dispersive solvent) into the sample solution. After phase separation by centrifugation, As(III), Se(IV) and Te(IV) preconcentrated in the organic phase were determined by ETV-ICP-MS. Total inorganic As, Se and Te were obtained by reducing As(V), Se(VI) and Te(VI) to As(III), Se(IV) and Te(IV) with L-cysteine, which were then subjected to the same DLLME-ETV-ICP-MS process. The concentration of As(V), Se(VI), Te(VI) were calculated by subtracting the concentration of As(III), Se(IV) and Te(IV) from the total inorganic As, Se and Te, respectively. The main factors affecting the microextraction efficiency and the vaporization behavior of target species were investigated in detail. Under the optimal conditions, the limits of detection were 2.5, 8.6 and 0.56 ng L(-1) for As(III), Se(IV) and Te(IV), respectively, with the relative standard deviations (n=7) of 8.5-9.7%. The developed method was applied to the speciation of inorganic As, Se and Te in Certified Reference Materials of GSBZ50004-88, GBW(E)080395 and GBW(E)080548 environmental waters, and the determined values are in good agreement with the certified values. The method was also successfully applied to the simultaneous speciation of inorganic As, Se and Te in different environmental water samples with the recoveries in the range of 86.3-107% for the spiked samples.
Related Materials