EMAIL THIS PAGE TO A FRIEND
Fish & shellfish immunology
Modulation of immunity and gut microbiota after dietary administration of alginate encapsulated Shewanella putrefaciens Pdp11 to gilthead seabream (Sparus aurata L.).
PMID 26003737
Abstract
The potential benefits of probiotics when administering to fish could improve aquaculture production. The objective of this study was to examine the modulation of immune status and gut microbiota of gilthead seabream (Sparus aurata L.) specimens by a probiotic when administered encapsulated. Commercial diet was enriched with Shewanella putrefaciens Pdp11 (SpPdp11, at a concentration of 10(8) cfu g(-1)) before being encapsulated in calcium alginate beads. Fish were fed non-supplemented (control) or supplemented diet for 4 weeks. After 1, 2 and 4 weeks the main humoral and cellular immune parameters were determined. Furthermore, gene expression profile of five immune relevant genes (il1β, bd, mhcIIα, ighm and tcrβ) was studied by qPCR in head kidney. On the other hand, intestinal microbiota of fish was analysed at 7 and 30 days by DGGE. Results demonstrated that administration of alginate encapsulated SpPdp11 has immunostimulant properties on humoral parameters (IgM level and serum peroxidase activity). Although no immunostimulant effects were detected on leucocyte activities, significant increases were detected in the level of mRNA of head-kidney leucocytes for mhcIIα and tcrβ after 4 weeks of feeding the encapsulated-probiotic diet. The administration of SpPdp11 encapsulated in alginate beads produced important changes in the DGGE patterns corresponding to the intestinal microbiota. Predominant bands related to lactic acid bacteria, such as Lactococcus and Lactobacillus strains, were sequenced from the DGGE patterns of fish fed the probiotic diet, whereas they were not sequenced from fish receiving the control diet. The convenience or not of probiotic encapsulation is discussed.