EMAIL THIS PAGE TO A FRIEND
Micron (Oxford, England : 1993)
Ultrastructure of antennal and posterior abdominal sensilla in Chlorophorus caragana females.
PMID 26004756
Abstract
Chlorophorus caragana Xie & Wang is a destructive wood-boring beetle that damages Caragana spp. bushes and is distributed in desert areas in north-west China. Using scanning electron microscopy and transmission electron microscopy, we observed the morphology and ultrastructure of antennal and posterior abdominal sensilla in C. caragana females, to discuss the putative functions of these sensilla in host location and oviposition behaviors. In total, seven types (24 subtypes) of sensilla were located on the antenna and posterior abdomen. On the antenna, there were Böhm's bristles (BB.); four subtypes of sensilla chaetica (Ch.1-Ch.4) characterized by non-porous surfaces and sensillum-lymph cavities without dendrites; two subtypes of sensilla trichodea (Tr.1 and Tr.2) with a tip pore and dendrites surrounded by dendritic sheaths; dome-shaped sensilla (Dom.) emerging from a deep cavity with one tip pore; four subtypes of sensilla basiconica (Ba.1-Ba.4) and one type of sensilla auricillica (Au.) with a porous cuticular surface and dendrites in the sensillum-lymph cavity; and one type of sensilla styloconica (Sty.) with grooves on the cuticular wall. On the posterior abdomen, there were four subtypes of sensilla chaetica (Ch.5-Ch.8); three subtypes of sensilla trichodea (Tr.3-Tr.5); and three subtypes of sensilla basiconica (Ba.5-Ba.7; Ba.5 had no groove in the cuticular wall, Ba.6 had one tip pore, and Ba.7 was located in a cuticular cavity). The antennal sensilla were believed to be mechanosensitive, chemosensitive, and sensitive to humidity and temperature, and to play roles in mating, host location and oviposition. The abdominal sensilla are believed to be related to oviposition behaviors.