EMAIL THIS PAGE TO A FRIEND
Journal of oral rehabilitation
In vitro acid decalcification of human surface enamel pretreated with toothpaste slurries.
PMID 2600715
Abstract
An in vitro caries-like challenge of human tooth enamel treated with inorganic fluoride (NaF) and organic fluoride (aliphatic and aromatic amine fluorides) toothpaste slurries has been investigated. The results revealed that the organic fluoride-containing toothpastes affected positively the resistance of tooth enamel to demineralization.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P66807
3-Pyridinemethanol, 98%
C6H7NO