EMAIL THIS PAGE TO A FRIEND
Journal of separation science
Quality control of processed Crataegi Fructus and its medicinal parts by ultra high performance liquid chromatography with electrospray ionization tandem mass spectrometry.
PMID 26009877
Abstract
Crataegi Fructus, an edible food, has been used as a traditional medicine to treat diseases for many years. There is substantial evidence that multiple constituents are responsible for the beneficial effects of Crataegi Fructus. To effectively control the quality of this herbal medicine, we developed an ultra high performance liquid chromatography with electrospray ionization tandem mass spectrometry protocol to simultaneously quantify ten compounds (chlorogenic acid, procyanidin B2, l-epicatechin, glucosylvitexin, vitexin-2-O-rhamnoside, vitexin, rutin, hyperoside, isoquercitrin, and quercetin) in Crataegi Fructus. Multiple-reaction monitoring was used for the quantification in the negative mode for 8 min. This proposed method is simple, reliable, sensitive, and specific. Further, the quantification parameters, including linearity, limit of detection, limit of quantification, precision, reproducibility, stability, and accuracy were optimized. The quality of the processed samples of Crataegi Fructus was evaluated using this method. Additionally, the method was successfully used to distinguish the medicinal components, including peel, kernel, and flesh. The data described in this study offer valuable information for the quality control and proper use of Crataegi Fructus.