EMAIL THIS PAGE TO A FRIEND
Oncotarget
The novel proteasome inhibitor carfilzomib activates and enhances extrinsic apoptosis involving stabilization of death receptor 5.
PMID 26009898
Abstract
Carfilzomib (CFZ) is a second generation proteasome inhibitor approved for the treatment of patients with multiple myeloma. It induces apoptosis in human cancer cells; but the underlying mechanisms remain undefined. In the present study, we show that CFZ decreases the survival of several human cancer cell lines and induces apoptosis. Induction of apoptosis by CFZ occurs, at least in part, due to activation of the extrinsic apoptotic pathway, since FADD deficiency protected cancer cells from undergoing apoptosis. CFZ increased total and cell surface levels of DR5 in different cancer cell lines; accordingly it enhanced TRAIL-induced apoptosis. DR5 deficiency protected cancer cells from induction of apoptosis by CFZ either alone or in combination with TRAIL. These data together convincingly demonstrate that DR5 upregulation is a critical mechanism accounting for CFZ-induced apoptosis and enhancement of TRAIL-induced apoptosis. CFZ inhibited the degradation of DR5, suggesting that DR5 stabilization contributes to CFZ-induced DR5 upregulation. In summary, the present study highlights the important role of DR5 upregulation in CFZ-induced apoptosis and enhancement of TRAIL-induced apoptosis in human cancer cells.