EMAIL THIS PAGE TO A FRIEND
Journal of clinical pharmacy and therapeutics
Celecoxib for the treatment of mild-to-moderate depression due to acute brucellosis: a double-blind, placebo-controlled, randomized trial.
PMID 26009929
Abstract
Depression is a debilitating complication of brucellosis and how best to treat this is a matter of debate. Inflammatory processes are involved in the pathogenesis of both brucellosis and depression. Therefore, we hypothesized that celecoxib could be beneficial for the treatment of depression due to brucellosis. Forty outpatients with depression due to brucellosis with a Hamilton Depression Rating Scale score (HDRS) <19 participated in a randomized, double-blind, placebo-controlled trial and underwent 8 weeks of treatment with either celecoxib (200 mg bid) or placebo as an adjunctive to antibiotic therapy. Patients were evaluated using HDRS at baseline and weeks 4 and 8. Repeated-measures analysis demonstrated significant effect for time × treatment interaction on the HDRS score [F (1·43, 57·41) = 37·22, P < 0·001]. Significantly greater response to treatment occurred in the celecoxib group than in the placebo group at the study end [10 patients (50%) vs. no patient (0%), respectively, P < 0·001]. No serious adverse event was observed. Celecoxib is a safe and effective treatment for depression due to brucellosis when compared with placebo.