EMAIL THIS PAGE TO A FRIEND
Epilepsia
Ketogenic diet exhibits anti-inflammatory properties.
PMID 26011473
Abstract
The ketogenic diet (KD) is an established treatment for refractory epilepsy, including some inflammation-induced epileptic encephalopathies. In a lipopolysaccharide (LPS)-induced fever model in rats, we found that animals given the KD for 14 days showed less fever and lower proinflammatory cytokine levels than control animals. However, KD rats exhibited a decrease in circulating levels of arachidonic acid and long-chain n-3 polyunsaturated fatty acids (PUFAs), suggesting that the anti-inflammatory effect of KD was probably not due to an increase in anti-inflammatory n-3 PUFA derivatives. These properties might be of interest in some conditions such as fever-induced refractory epileptic encephalopathy in school-aged children.