EMAIL THIS PAGE TO A FRIEND
ChemSusChem
Hierarchically Structured Hole Transport Layers of Spiro-OMeTAD and Multiwalled Carbon Nanotubes for Perovskite Solar Cells.
PMID 26013428
Abstract
The low electrical conductivity of spiro-OMeTAD hole transport layers impedes further enhancements of the power conversion efficiency (PCE) of perovskite solar cells. We embedded multiwalled carbon nanotubes (MWNTs) in spiro-OMeTAD (spiro-OMeTAD/MWNTs) to increase carrier mobility and conductivity. However, direct electrical contact between CH3 NH3 PbI3 and the MWNTs created pathways for undesirable back-electron transfer, owing to the large work function of MWNTs, limiting enhancements of the PCE. A hierarchical structure of pure spiro-OMeTAD and spiro-OMeTAD/MWNTs was designed to block back-electron transfer and fully exploit the enhanced charge transport of spiro-OMeTAD/MWNTs. The enhanced fill factor, short-circuit current density, open-circuit voltage, and PCE (15.1 %) were achieved by using this hierarchical hole transport layer structure (MWNT concentration=2 wt %). The perovskite solar cells were fabricated by a low-temperature solution process, further decreasing their per-Watt cost.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
284513
Chlorobenzene, anhydrous, 99.8%
C6H5Cl