EMAIL THIS PAGE TO A FRIEND
Molecular therapy. Methods & clinical development
AAV8-mediated Sirt1 gene transfer to the liver prevents high carbohydrate diet-induced nonalcoholic fatty liver disease.
PMID 26015978