EMAIL THIS PAGE TO A FRIEND
Scientific reports
Direct identification of on-bead peptides using surface-enhanced Raman spectroscopic barcoding system for high-throughput bioanalysis.
PMID 26017924
Abstract
Recently, preparation and screening of compound libraries remain one of the most challenging tasks in drug discovery, biomarker detection, and biomolecular profiling processes. So far, several distinct encoding/decoding methods such as chemical encoding, graphical encoding, and optical encoding have been reported to identify those libraries. In this paper, a simple and efficient surface-enhanced Raman spectroscopic (SERS) barcoding method using highly sensitive SERS nanoparticles (SERS ID) is presented. The 44 kinds of SERS IDs were able to generate simple codes and could possibly generate more than one million kinds of codes by incorporating combinations of different SERS IDs. The barcoding method exhibited high stability and reliability under bioassay conditions. The SERS ID encoding based screening platform can identify the peptide ligand on the bead and also quantify its binding affinity for specific protein. We believe that our SERS barcoding technology is a promising method in the screening of one-bead-one-compound (OBOC) libraries for drug discovery.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
275425
2,5-Dimethoxythiophenol, 98%
C8H10O2S
541850
2-Amino-4-chlorobenzenethiol, 96%
C6H6ClNS
242659
2-Bromothiophenol, 97%
C6H5BrS
247146
2-Chlorothiophenol, 99%
C6H5ClS
275379
2-Fluorothiophenol, 97%
C6H5FS
301507
2-Mercapto-1-methylimidazole, ≥99%
C4H6N2S
642096
2-Mercapto-6-methylpyridine, 97%
C6H7NS
M3205
2-Mercaptobenzimidazole, 98%
C7H6N2S
129623
2-Mercaptopyrimidine, 98%
C4H4N2S
270849
2-Naphthalenethiol, 99%
C10H8S
116270
2-Quinolinethiol, 97%
C9H7NS
M6204
2-Thiazoline-2-thiol, 98%
C3H5NS2
T7750
2-Thiouracil, ≥99%
C4H4N2OS
662518
3,4-Dimethoxythiophenol, 97%
C8H10O2S
269689
3,4-Dimethylbenzenethiol, 98%
C8H10S
522929
3,5-Dimethylbenzenethiol, 90%
C8H10S
140260
3-Amino-1,2,4-triazole-5-thiol, 95%
C2H4N4S
157163
3-Cyanobenzoic acid, 98%
C8H5NO2
451436
3-Mercaptobenzoic acid, 95%
C7H6O2S
289426
4,4′−Dipyridyl, 98%
C10H8N2
422967
4-Aminothiophenol, ≥97.0% (GC)
C6H7NS
B81956
4-Bromothiophenol, 95%
C6H5BrS
125237
4-Chlorothiophenol, 97%
C6H5ClS
F15315
4-Fluorothiophenol, 98%
C6H5FS
706329
4-Mercaptobenzoic acid, 99%
C7H6O2S
662534
4-Mercaptobenzoic acid, technical grade, 90%
C7H6O2S
109525
4-Methoxythiophenol, 97%
C7H8OS
T28525
4-Methylbenzenethiol, 98%
C7H8S
N21022
4-Nitrophenyl disulfide
C12H8N2O4S2
438561
5-(4-Pyridyl)-1,3,4-oxadiazole-2-thiol, 97%
C7H5N3OS