EMAIL THIS PAGE TO A FRIEND
Human molecular genetics
Transcriptome outlier analysis implicates schizophrenia susceptibility genes and enriches putatively functional rare genetic variants.
PMID 26022996
Abstract
We searched a gene expression dataset comprised of 634 schizophrenia (SZ) cases and 713 controls for expression outliers (i.e., extreme tails of the distribution of transcript expression values) with SZ cases overrepresented compared with controls. These outlier genes were enriched for brain expression and for genes known to be associated with neurodevelopmental disorders. SZ cases showed higher outlier burden (i.e., total outlier events per subject) than controls for genes within copy number variants (CNVs) associated with SZ or neurodevelopmental disorders. Outlier genes were enriched for CNVs and for rare putative regulatory variants, but this only explained a small proportion of the outlier subjects, highlighting the underlying presence of additional genetic and potentially, epigenetic mechanisms.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A9883
ADA, ≥98% (titration)
C6H10N2O5
A8074
ADA, BioXtra, ≥98% (titration)
C6H10N2O5