EMAIL THIS PAGE TO A FRIEND
Journal of liposome research
Effect of lipid composition on incorporation of trastuzumab-PEG-lipid into nanoliposomes by post-insertion method: physicochemical and cellular characterization.
PMID 26023889
Abstract
Anti-HER2 immunoliposomes are promising nanotechnology based systems for active targeting of breast tumors, which depends on the amount of incorporated antibody. In this work, we investigated the possible effect of lipid composition on the incorporation of trastuzumab-PEG-PE micelles into nanoliposomes and on their subsequent specific cellular targeting. Trastuzumab (anti-HER2 monoclonal antibody) was monothiolated and conjugated to maleimide-PEG-PE micelles. Liposomes of different lipid compositions were prepared by the thin layer hydration. Trastuzumab-PEG-PE micelles were incorporated into the liposomes by the post-insertion method. The percentage of lipid mixing was determined based on fluorescence resonance energy transfer. Cellular binding and uptake of rhodamine-labeled immunoliposomes were studied in SKBR-3 (HER2(+++)) and MCF-7 (HER2(+)) cells. Also, antitumor cell activity of the immunoliposomes was compared to free trastuzumab and the liposomes. The lipid mixing of trastuzumab-PEG-PE micelles depended on the liposome composition. The immunoliposomes containing DPPC, cholesterol and PEG-PE showed prominent lipid mixing. The lipid mixing was consistent with the cell binding results which showed an efficient and specific binding of the immunoliposomes to SKBR-3 cells. Antitumor cell activity of the immunoliposomes in SKBR-3, unlike MCF-7 cells, depended on the content of trastuzumab. Cholesterol and PEG-PE in the liposome composition are prerequisites for a successful lipid mixing due to their ability to facilitate fusion. The higher lipid mixing results in higher antibody incorporation and consequently higher targeted cell binding. The lipid mixing depends on the liposome composition, which reflects targeted cell binding of the immunoliposomes.