EMAIL THIS PAGE TO A FRIEND
Blood
High-density preculture of PBMCs restores defective sensitivity of circulating CD8 T cells to virus- and tumor-derived antigens.
PMID 26024876
Abstract
Peripheral blood mononuclear cells (PBMCs) are the only source of human lymphoid cells routinely available for immunomonitoring of T-cell responses to microbial and tumor-associated antigens. However, previous work in mice and humans had indicated that CD4 T cells transiently lose antigen sensitivity when cellular contacts are lost (eg, by entering the circulation). Using the simple and robust protocol for resetting T cells to original reactivity (RESTORE; ie, preculturing PBMCs for 2 days at a high cell density before initiation of antigenic stimulation), we show that CD8 T-cell responses to viral and tumor-associated antigens are greatly underestimated in blood, and sometimes even remain undetected, if conventional, unprocessed PBMC cultures are used. The latter finding is particularly striking with regard to the appearance of Wilms tumor 1 protein-specific CD8 T-cell responses in leukemia patients after allogeneic bone marrow transplantation. The dramatic increase in antigen sensitivity of "restored" CD8 T cells is associated with phosphorylation of proximal T-cell receptor signaling components, and with the upregulation of genes involved in aerobic glycolysis, thereby increasing T-cell functionality. The RESTORE protocol permits a more meaningful monitoring of CD8 memory T-cell responses to viral infections and tumors and vaccination success. Furthermore, when generating T-cell lines for adoptive T-cell therapy, it avoids the loss of those clones, which strictly depend on the primed status conferred by cellular interactions in the tissue context for their initial reactivation by antigen. The data reported in this article have been deposited in the Gene Expression Omnibus database (accession number GSE63430).