EMAIL THIS PAGE TO A FRIEND
Antonie van Leeuwenhoek
Description of Geodermatophilus bullaregiensis sp. nov.
PMID 26025305
Abstract
The taxonomic position of an aerobic actinobacterial strain, BMG841(T), isolated from the Bulla Regia monument (Tunisia) and exhibiting a high resistance to gamma-radiation (D10 ~9 kGy) was determined using polyphasic approach. The optimal growth range was found to be 25-35 °C at pH of 7.0-8.5. The strain was observed to form black dry colonies. Chemotaxonomic characteristics of the isolate showed a cell wall type III, with galactose and glucose as diagnostic sugars; phosphatidylcholine, phosphatidylinositol, diphosphatidylglycerol, phosphatidylethanolamine and an unidentified glycolipid as main polar lipids; and MK-9(H4) as the predominant menaquinone. The major cellular fatty acids were identified as iso-C16:0 and iso-C15:0. Phylogenetic analysis indicated that strain BMG841(T) represents a novel member of the genus Geodermatophilus with high 16S rRNA gene sequence identity with Geodermatophilus saharensis (98.28 %). Based on phylogenetic and phenotypic analysis, strain BMG841(T) is proposed as the type strain (=DSM 46841(T) = CECT 8821(T)) of a novel species, Geodermatophilus bullaregiensis.