EMAIL THIS PAGE TO A FRIEND
Toxicology in vitro : an international journal published in association with BIBRA
Cyto- and genotoxic effects of metallic nanoparticles in untransformed human fibroblast.
PMID 26028148
Abstract
Metallic nanoparticles such as silver (Ag), cerium dioxide (CeO2) and titanium dioxide (TiO2) are produced at a large scale and included in many consumer products. It is well known that most metallic NPs are toxic to humans which raise concerns about these engineered particles. Various studies have already been published on the subject, however, almost all of these studies have been conducted in cancer or transformed cell lines. In this work we performed a comparative evaluation of these metallic NPs on normal untransformed human fibroblasts (GM07492) detecting cyto- and geno-toxic responses after exposure to these NPs. Our results showed that all three metallic NPs were able to cross the plasma membrane and were mainly found in endocytic vesicles. The Ag and TiO2 NPs affected mitochondrial enzymatic activity (XTT), increased DNA fragmentation, oxidative damage (Comet assay) and induced cell death mainly by the apoptotic pathway. Ag NPs increased GADD45α transcript levels and the phosphorylation of proteins γH2AX. Transient genotoxicity was also observed from exposure to CeO2 NPs while TiO2 NPs showed no increase in DNA damage at sub-cytotoxic concentrations. In comparison, Ag NPs were found to be the most cyto-genotoxic NPs to fibroblasts. Thus, these results support the use of normal fibroblast as a more informative tool to detect the mechanisms of action induced by metallic NPs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
129925
Methyl methanesulfonate, 99%
C2H6O3S
677469
Titanium(IV) oxide, contains 1% Mn as dopant, nanopowder, <100 nm particle size (BET), ≥97%
O2Ti
718467
Titanium(IV) oxide, nanopowder, 21 nm primary particle size (TEM), ≥99.5% trace metals basis
O2Ti
774510
Titanium(IV) oxide, nanowires, diam. × L ~100 nm × 10 μm
O2Ti
774529
Titanium(IV) oxide, nanowires, diam. × L ~10 nm × 10 μm
O2Ti
637254
Titanium(IV) oxide, anatase, nanopowder, <25 nm particle size, 99.7% trace metals basis
O2Ti
232033
Titanium(IV) oxide, anatase, powder, 99.8% trace metals basis
O2Ti
248576
Titanium(IV) oxide, anatase, powder, −325 mesh, ≥99% trace metals basis
O2Ti
634662
Titanium(IV) oxide, mixture of rutile and anatase, nanopowder, <100 nm particle size (BET), 99.5% trace metals basis
O2Ti
700355
Titanium(IV) oxide, mixture of rutile and anatase, nanoparticle, <250 nm particle size (DLS), paste, 53-57 wt. % in diethylene glycol monobutyl ether/ethylene glycol, 99.9% trace metals basis
O2Ti
700347
Titanium(IV) oxide, mixture of rutile and anatase, nanoparticles, <150 nm particle size (volume distribution, DLS), dispersion, 33-37 wt. % in H2O, 99.5% trace metals basis
O2Ti
700339
Titanium(IV) oxide, mixture of rutile and anatase, nanoparticles, <100 nm particle size, dispersion, 48-52 wt. % in xylene, 99.9% trace metals basis
O2Ti
204730
Titanium(IV) oxide, rutile, 99.995% trace metals basis
O2Ti
204757
Titanium(IV) oxide, rutile, ≥99.98% trace metals basis
O2Ti
635057
Titanium(IV) oxide, rutile, <001>, (single crystal substrate), 99.99% trace metals basis, L × W × thickness 10 mm × 10 mm × 0.5 mm
O2Ti
635049
Titanium(IV) oxide, rutile, <100>, (single crystal substrate), 99.99% trace metals basis, L × W × thickness 10 mm × 10 mm × 0.5 mm
O2Ti
635065
Titanium(IV) oxide, rutile, <110>, single crystal substrate, 99.99% trace metals basis, L × W × thickness 10 mm × 10 mm × 0.5 mm
O2Ti
637262
Titanium(IV) oxide, rutile, nanopowder, <100 nm particle size, 99.5% trace metals basis
O2Ti
224227
Titanium(IV) oxide, rutile, powder, <5 μm, ≥99.9% trace metals basis
O2Ti