EMAIL THIS PAGE TO A FRIEND
Environmental Science and Pollution Research International
Chemometric and environmental assessment of arsenic, antimony, and chromium speciation form occurrence in a water reservoir subjected to thermal anthropopressure.
PMID 26028350