Microscopy research and technique
Three-dimensional co-culture of hepatic progenitor cells and mesenchymal stem cells in vitro and in vivo.
PMID 26031767
Here we co-cultured hepatic progenitor cells (HPCs) and mesenchymal stem cells (MSCs) to investigate whether the co-culture environments could increase hepatocytes form. Three-dimensional (3D) co-culture model of HPCs and MSCs was developed and morphological features of cells were continuously observed. Hepatocyte specific markers Pou5f1/Oct4, AFP, CK-18 and Alb were analyzed to confirm the differentiation of HPCs. The mRNA expression of CK-18 and Alb was analyzed by RT-PCR to investigate the influence of co-culture model to the terminal differentiation process of mature hepatocytes. The functional properties of hepatocyte-like cells were detected by continuously monitoring the albumin secretion using Gaussia luciferase assays. Scaffolds with HPCs and MSCs were implanted into nude mouse subcutaneously to set up the in vivo co-culture model. Although two groups formed smooth spheroids and high expressed of CK-18 and Alb, hybrid spheroids had more regular structures and higher cell density. CK-18 and Alb mRNA were at a relatively higher expression level in co-culture system during the whole cultivation time (P < 0.05). Albumin secretion rates in the hybrid spheroids had been consistently higher than that in the mono-culture spheroids (P < 0.05). In vivo, the hepatocyte-like cells were consistent with the morphological features of mature hepatocytes and more well-differentiated hepatocyte-like cells were observed in the co-culture group. HPCs and MSCs co-culture system is an efficient way to form well-differentiated hepatocyte-like cells, hence, may be helpful to the cell therapy of hepatic tissues and alleviate the problem of hepatocytes shortage.
Related Materials
Product #
Molecular Formula
Add to Cart
9-cis-Retinoic acid, ≥98% (HPLC)