EMAIL THIS PAGE TO A FRIEND
The British journal of dermatology
Levels and function of regulatory T cells in patients with polymorphic light eruption: relation to photohardening.
PMID 26032202
Abstract
We hypothesized that regulatory T cells (Tregs) are involved in the immunological abnormalities seen in patients with polymorphic light eruption (PLE). To investigate the number and suppressive function of peripheral Tregs in patients with PLE compared with healthy controls. Blood sampling was done in 30 patients with PLE [seeking or not seekingxa0311-nm ultraviolet (UV)B photohardening] as well as 19 healthy controls at two time points: TP1, March to June (before phototherapy); and TP2,xa0May to August (after phototherapy). We compared the number of CD4(+) CD25(high) CD127(-) FoxP3(+) Tregs by flow cytometry and their function by assessing FoxP3 mRNA levels and effector T cell/Treg suppression assays. Tregs isolated from healthy controls significantly suppressed the proliferation of effector T cells at TP1 by 68% (Pxa0=xa00·0156). In contrast, Tregs from patients with PLE entirely lacked the capacity to suppress effector T-cell proliferation at that time point. The medical photohardening seen in 23 patients with PLE resulted in a significant increase in the median percentage of circulating Tregs [both as a proportion of all lymphocytes; 65 6% increase (Pxa0=xa00·0049), and as a proportion of CD4(+) T cells; 32.5% increase (Pxa0=xa00·0049)]. This was accompanied by an increase in the expression of FoxP3 mRNA (Pxa0=xa00·0083) and relative immunosuppressive function of Tregs (Pxa0=xa00·083) comparing the two time points in representative subsets of patients with healthy controls tested. Seven patients with PLE not receiving 311-nm UVB also exhibited an increase in the number of Tregs but this was not statistically significant. No significant differences in Treg numbers were observed in healthy subjects between the two time points. An impaired Treg function is likely to play a role in PLE pathogenesis. A UV-induced increase in the number of Tregs (either naturally or therapeutically) may be a compensatory mechanism by which the immune system counteracts the susceptibility to PLE.