EMAIL THIS PAGE TO A FRIEND
Veterinary surgery : VS
Single-Incision Laparoscopic Sterilization of the Cheetah (Acinonyx jubatus).
PMID 26032294
Abstract
To describe laparoscopic ovariectomy and salpingectomy in the cheetah (Acinonyx jubatus) using single-incision laparoscopic surgery (SILS). Prospective cohort. Female cheetahs (Acinonyx jubatus) (n = 21). Cheetahs were randomly divided to receive either ovariectomy (n = 11) or salpingectomy (n = 10). The use and complications of a SILS port was evaluated in all of cheetahs. Surgery duration and insufflation volumes of carbon dioxide (CO2 ) were recorded and compared across procedures. Laparoscopic ovariectomy and salpingectomy were performed without complications using a SILS port. The poorly-developed mesosalpinx and ovarian bursa facilitated access to the uterine tube for salpingectomy in the cheetah. The median surgery duration for ovariectomy was 24 minutes (interquartile range 3) and for salpingectomy was 19.5 minutes (interquartile range 3) (P = .005). The median volume of CO2 used for ovariectomy was 11.25 L (interquartile range 3.08) and for salpingectomy was 4.90 L (interquartile range 2.52), (P = .001) CONCLUSIONS: Laparoscopic ovariectomy and salpingectomy can be performed in the cheetah using SILS without perioperative complications. Salpingectomy is faster than ovariectomy and requires less total CO2 for insufflation.