EMAIL THIS PAGE TO A FRIEND
Scientific reports
Tumor-specific Th2 responses inhibit growth of CT26 colon-cancer cells in mice via converting intratumor regulatory T cells to Th9 cells.
PMID 26035423
Abstract
The abnormality of immune regulation plays a critical role in the pathogenesis of cancer; the underlying mechanism has not been fully understood yet. This study aims to investigate the role of cancer specific T helper (Th)2 response in the inhibition of colon cancer (Cca) cell growth. The results showed that with Cca cell (CT26 cell) extracts as an antigen, the Cca-extract specific Th2 response was induced in the Cca-bearing mice. The Cca mass size was significantly reduced, or radically disappeared (5 out of 10; or 50%); the survival rate was markedly improved in mice immunized with Cca-extract, but not in those immunized with another tumor cell (U87 cell) extracts or to bovine serum albumin. The immunization with Cca-extract also induced Cca cell apoptosis and converted the intra-Cca Tregs to T helper (Th) 9 cells. In conclusion, Cca-specific Th2 responses inhibit Cca growth in a mouse model via inducing Cca cell apoptosis and converting intra-Cca Tregs to Th9 cells.