EMAIL THIS PAGE TO A FRIEND
European Journal of Pharmaceutics and Biopharmaceutics
Formulation to target delivery to the ciliary body and choroid via the suprachoroidal space of the eye using microneedles.
PMID 26036448