EMAIL THIS PAGE TO A FRIEND
The New phytologist
Grassland species differentially regulate proline concentrations under future climate conditions: an integrated biochemical and modelling approach.
PMID 26037253