EMAIL THIS PAGE TO A FRIEND
The Journal of allergy and clinical immunology
Peanut defensins: Novel allergens isolated from lipophilic peanut extract.
PMID 26037551
Abstract
Peanut is one of the most hazardous sources of food allergens. Unknown allergens are still hidden in the complex lipophilic matrix. These allergens need to be discovered to allow estimation of the allergenic risk for patients with peanut allergy and to further improve diagnostic measures. We performed detection, isolation, and characterization of novel peanut allergens from lipophilic peanut extract. Extraction of roasted peanuts were performed under defined extraction conditions and examined by means of 2-dimensional PAGE. Subsequently, chromatographic methods were adapted to isolate low-molecular-weight components. Proteins were studied by using SDS-PAGE and immunoblotting with sera from patients with peanut allergy. For allergen identification protein sequencing, homology search and mass spectrometry were applied. Functional characterization for allergenicity was performed by using the basophil activation assay and for antimicrobial activity by using inhibition assays of different bacteria and fungi. IgE-reactive proteins of 12, 11, and 10 kDa were first detected after chloroform/methanol extraction in the flow through of hydrophobic interaction chromatography. The proteins were able to activate basophils of patients with peanut allergy. N-terminal sequencing and homology search in the expressed sequence tag database identified the allergens as peanut defensins, which was confirmed by using mass spectrometry. On microbial cell cultures, the peanut defensins showed inhibitory effects on the mold strains of the genera Cladosporium and Alternaria but none on bacteria. We identified defensins as novel peanut allergens (Ara h 12 and Ara h 13) that react in particular with IgE of patients with severe peanut allergy. Their antimicrobial activity is solely antifungal.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F3506
N-Formyl-Met-Leu-Phe, ≥97% (HPLC)
C21H31N3O5S
47729
N-Formyl-Met-Leu-Phe, BioXtra, ≥99.0% (TLC)
C21H31N3O5S