EMAIL THIS PAGE TO A FRIEND
Stem cell research & therapy
99m-Technetium binding site in bone marrow mononuclear cells.
PMID 26041023
Abstract
The increasing interest in 99m-technetium ((99m)Tc)-labeled stem cells encouraged us to study the (99m)Tc binding sites in stem cell compartments. Bone marrow mononuclear cells were collected from femurs and tibia of rats. Cells were labeled with (99m)Tc by a direct method, in which reduced molecules react with (99m)Tc with the use of chelating agents, and lysed carefully in an ultrasonic apparatus. The organelles were separated by means of differential centrifugation. At the end of this procedure, supernatants and pellets were counted, and the percentages of radioactivity (in megabecquerels) bound to the different cellular fractions were determined. Percentages were calculated by dividing the radioactivity in each fraction by total radioactivity in the sample. The pellets were separated and characterized by their morphology on electron microscopy. The labeling procedure did not affect viability of bone marrow mononuclear cells. Radioactivity distributions in bone marrow mononuclear cell organelles, obtained in five independent experiments, were approximately 38.5 % in the nuclei-rich fraction, 5.3 % in the mitochondria-rich fraction, 2.2 % in microsomes, and 54 % in the cytosol. Our results showed that most of the radioactivity remained in the cytosol; therefore, this is an intracellular labeling procedure that has ribosomes unbound to membrane and soluble molecules as targets. However, approximately 39 % of the radioactivity remained bound to the nuclei-rich fraction. To confirm that cell disruption and organelle separation were efficient, transmission electron microscopy assays of all pellets were performed. Our results showed that most of the radioactivity was present in the cytosol fraction. More studies to elucidate the mechanisms involved in the cellular uptake of (99m)Tc in bone marrow cells are ongoing.