EMAIL THIS PAGE TO A FRIEND
Science translational medicine
Regulator of G protein signaling 5 is a determinant of gestational hypertension and preeclampsia.
PMID 26041705
Abstract
Preeclampsia is a systemic vascular disorder of pregnancy and is associated with increased sensitivity to angiotensin II (AngII) and hypertension. The cause of preeclampsia remains unknown. We identified the role of regulator of G protein (heterotrimeric guanine nucleotide-binding protein) signaling 5 (RGS5) in blood pressure regulation during pregnancy and preeclampsia. RGS5 expression in human myometrial vessels is markedly suppressed in gestational hypertension and/or preeclampsia. In pregnant RGS5-deficient mice, reduced vascular RGS5 expression causes gestational hypertension by enhancing vascular sensitivity to AngII. Further challenge by increasing AngII results in preeclampsia-like symptoms, namely, more severe hypertension, proteinuria, placental pathology, and reduced birth weight. In pregnant heterozygote null mice, treatment with peroxisome proliferator-activated receptor (PPAR) agonists normalizes vascular function and blood pressure through effects on RGS5. These findings highlight a key role of RGS5 at the interface between AngII and PPAR signaling. Because preeclampsia is refractory to current standard therapies, our study opens an unrecognized and urgently needed opportunity for treatment of gestational hypertension and preeclampsia.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A10809
4′-Hydroxy-3′-methoxyacetophenone, 98%
C9H10O3
W508454
Acetovanillone, ≥98%, FG
C9H10O3