EMAIL THIS PAGE TO A FRIEND
International journal for parasitology. Drugs and drug resistance
In vitro and in vivo assessment of the effect of antiprotozoal compounds isolated from Psoralea corylifolia against Ichthyophthirius multifiliis in fish.
PMID 26042195
Abstract
Ichthyophthirius multifiliis, an external fish parasite, often causes significant economic damage to the aquaculture industry. Since the use of malachite green was banned, the search of alternative substance to control I. multifiliis infections becomes stringent. In present study, in vitro and in vivo anti-ich efficacies of isopsoralen and psoralidin, two active compounds isolated from methanol extract of Psoralea corylifolia by bioassay-guided fractionation based on the efficacy of anti-ich encysted tomonts, were evaluated. In vitro antiprotozoal efficacy of psoralidin is much better than that of isopsoralen. Psoralidin can kill all theronts at concentrations of 0.8 mg/L or more during 4 h exposure; and terminate reproduction of I. multifiliis post 6 h exposure of protomonts to 0.9 mg/L and encysted tomonts to 1.2 mg/L. In vivo trials showed that 5 h exposure of infected fish to 2.5 mg/L of psoralidin significantly reduced the number of theronts released from tomonts. Furthermore, we observed that a part of protomonts, collected from infected fish post treatment, presented characteristic morphological changes of apoptosis after staining with Annexin V-EGFP/propidium iodide, indicating the possible mechanism of psoralidin against I. multifiliis trophont in situ. On the basis of these results, psoralidin can be used as a potential lead compound for the development of commercial drug against I. multifiliis.