EMAIL THIS PAGE TO A FRIEND
Bosnian journal of basic medical sciences
Immunization with 3-oxododecanoyl-L-homoserine lactone-r-PcrV conjugate enhances survival of mice against lethal burn infections caused by Pseudomonas aeruginosa.
PMID 26042508
Abstract
Quorum Sensing and type III secretion system play an important role in the virulence of Pseudomonas (P.) aeruginosa in burn wound infections. We aimed to explore the feasibility of using 3-oxo-C₁₂-HSL-r-PcrV conjugate as a candidate vaccine against P. aeruginosa caused infections. 3-oxo-C₁₂-HSL-r-PcrV conjugate was prepared and used for immunization of mice (10 μg, subcutaneous, three times, at 2-week intervals). Mice were divided into five groups: I: PcrV; II: 3-oxo-C₁₂-HSL-r-PcrV (10 μg); III: 3-oxo-C₁₂-HSL-r-PcrV (20 μg); IV: 3-oxo-C₁₂-HSL; and V: PBS receiving groups. After each shot of immunization, total and isotype antibody responses against corresponding antigen were measured to determine the immunization efficacy. One month after the last immunization, all groups were burned and challenged subeschar with P. aeruginosa PAO1. Survival rate and bacterial quantity in the skin and internal organs (liver and spleen) were evaluated 25-hr after burn infection. Immunization with 3-oxo-C₁₂-HSL-r-PcrV significantly increased total IgG and specific subclass antibodies (IgG₁, IgG₂a, IgG₂b, and IgM) in the serum of the groups II and III compared to the control group (p<0.001). While all the control mice (PBS injected group) died within 2 days after bacterial challenge, 64% of the group I, 78% of group II, and 86% of group III, survived within 14 days after challenge. Interestingly, bacterial burden in the liver and spleen of 3-oxo-C₁₂-HSL-r-PcrV injected group (III) was significantly lower than the control group (p<0.001). The present study proposed two-component vaccine to inhibit Pseudomonas infections in burned mouse.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O9139
N-(3-Oxododecanoyl)-L-homoserine lactone, quorum sensing signaling molecule
C16H27NO4