EMAIL THIS PAGE TO A FRIEND
Archives of oral biology
Effect of Capparis spinosa extract on sutural ossification: A stereological study.
PMID 26043444
Abstract
To investigate the effects of systemically administered Capparis spinosa extract (CSE) on expanded sutures in rats via three dimensionally morphometric method (stereological method). Thirty-two Wistar rats were used. Subjects were divided into four groups, each with eight rats. Orthopaedic expansion force was applied for 5 days to maxillary incisors by attaching springs. Control-1 and CSE-1 waited 1 week for consolidation, and Control-2 and CSE-2 waited 2 weeks for consolidation. After the consolidation period, the subjects were sacrificed. Stereological examination was performed to determine the volume and area of new bone, connective tissue, and capillaries. New bone area, new bone volume, connective tissue space, and connective tissue volume were statistically different in CSE-1 compared to Control-1. But there were no statistically difference between CSE-2 and Control_2. In terms of the volume of blood vessels and vascular area, there were no statistically significant differences when comparing Groups CSE-1 and Control-1 or CSE-2 and Control-2. Systemic use of CSE accelerated fastened osteoblastic activity in the early period.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T1019
(±)-Thiopental
C11H18N2O2S