EMAIL THIS PAGE TO A FRIEND
Apoptosis : an international journal on programmed cell death
PUMA-mediated mitochondrial apoptotic disruption by hypoxic postconditioning.
PMID 26043893
Abstract
Postconditioning can reduce ischemia-reperfusion (I/R)-induced cardiomyocyte apoptosis by targeting mitochondria. p53 upregulated modulator of apoptosis (PUMA) is involved in lethal I/R injury. Here, we hypothesized that postconditioning might inhibit mitochondrial pathway-mediated cardiomyocyte apoptosis by controlling PUMA expression. The cultured neonatal rat cardiomyocytes underwent 3xa0h of hypoxia and 3xa0h of reoxygenation. Postconditioning consisted of three cycles of 5xa0min reoxygenation and 5xa0min hypoxia after prolonged hypoxia. Hypoxic postconditioning reduced the levels of PUMA mRNA and protein. Concomitantly, the loss of mitochondrial membrane potential, cytochrome c release and caspase-3 activation were decreased significantly by postconditioning. Overexpression of PUMA increased greatly not only the number of apoptotic cardiomyocytes, but also the collapse of mitochondrial membrane potential, cytochrome c release and caspase-3 activation under postconditioning condition. The data suggest that reduction of PUMA expression mediates the endogenous cardioprotective mechanisms of postconditioning by disrupting mitochondrial apoptotic pathway.