EMAIL THIS PAGE TO A FRIEND
Biological trace element research
Activity of Selected Antioxidant Enzymes, Selenium Content and Fatty Acid Composition in the Liver of the Brown Hare (Lepus europaeus L.) in Relation to the Season of the Year.
PMID 26043915