EMAIL THIS PAGE TO A FRIEND
Environmental Science and Pollution Research International
Human health risk assessment of heavy metals in tropical fish and shellfish collected from the river Buriganga, Bangladesh.
PMID 26044144