EMAIL THIS PAGE TO A FRIEND
Small (Weinheim an der Bergstrasse, Germany)
Synthesis of Nanogels via Cell Membrane-Templated Polymerization.
PMID 26044721
Abstract
The synthesis of biomimetic hydrogel nanoparticles coated with a natural cell membrane is described. Compared to the existing strategy of wrapping cell membranes onto pre-formed nanoparticle substrates, this new approach forms the cell membrane-derived vesicles first, followed by growing nanoparticle cores in situ. It adds significant controllability over the nanoparticle properties and opens unique opportunities for a broad range of biomedical applications.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
163945
4-Amino-TEMPO, free radical, 97%
C9H19N2O