EMAIL THIS PAGE TO A FRIEND
International journal of clinical and experimental pathology
miR-200a/miR-141 and miR-205 upregulation might be associated with hormone receptor status and prognosis in endometrial carcinomas.
PMID 26045795
Abstract
The aim of this study was to compare the clinicopathological significance of miR-200a/miR-141 and miR-205 expression in endometrioid carcinomas (ECs) versus nonendometrioid carcinomas (NECs) and to assess their correlation with hormone receptor status. miR-200a/miR-141 and miR-205 expression in 154 endometrial cancers was determined by qRT-PCR. The status of estrogen and progesterone receptor (ER/PR) was assessed using immunohistochemistry. miR-200a/miR-141 and miR-205 increased significantly in ECs and in NECs. The expression level of miR-200a was significantly higher in NECs than in ECs (P=0.025). Furthermore, there was a trend that NECs with worse clinicopathological variables had a higher miR-200a expression, while an inverse trend existed in ECs. miR-205 upregulation occurred frequently in NECs without lymph node metastases (P=0.030), whereas such association was not present in ECs. Interestingly, In ECs, miR-200a/miR-141 upregulation occurred frequently in the hormone receptor positive subgroups than the negative subgroups (P<0.05). Similarly, the expression level of miR-205 was higher in the hormone receptor positive subgroups and the association between miR-205 and PR reached statistical significance (P=0.024). In contrast, in NECs, a negative correlation was found between miR-200a/miR-141 and ER or PR status. Meanwhile, in ECs, miR-200a upregulation correlated with prolonged survival in the ER positive subgroup (P=0.046), whereas an inverse trend existed in the ER negative subgroup. Our findings suggest that miR-200a/miR-141 and miR-205 increased significantly in ECs and in NECs. However, they might behave differently in ECs versus NECs. miR-200a/miR-141 and miR-205 might be associated with hormone receptor status in endometrial cancer and may possess prognostic impacts.