PloS one
Development of Sustained Release "NanoFDC (Fixed Dose Combination)" for Hypertension - An Experimental Study.
PMID 26047011
Abstract
The present study was planned to formulate, characterize and evaluate the pharmacokinetics of a novel "NanoFDC" comprising three commonly prescribed anti-hypertensive drugs, hydrochlorothiazide (a diuretic), candesartan (ARB) and amlodipine (a calcium channel blocker). The candidate drugs were loaded in Poly (DL-lactide-co-gycolide) (PLGA) by emulsion- diffusion-evaporation method. The formulations were evaluated for their size, morphology, drug loading and in vitro release individually. Single dose pharmacokinetic profiles of the nanoformulations alone and in combination, as a NanoFDC, were evaluated in Wistar rats. The candidate drugs encapsulated inside PLGA showed entrapment efficiencies ranging from 30%, 33.5% and 32% for hydrochlorothiazide, candesartan and amlodipine respectively. The nanoparticles ranged in size from 110 to 180 nm. In vitro release profile of the nanoformulation showed 100% release by day 6 in the physiological pH 7.4 set up with PBS (phosphate buffer saline) and by day 4-5 in the intestinal pH 1.2 and 8.0 set up SGF (simulated gastric fluid) and SIF (simulated intestinal fluid) respectively. In pharmacokinetic analysis a sustained-release for 6 days and significant increase in the mean residence time (MRT), as compared to the respective free drugs was noted [MRT of amlodipine, hydrochlorothiazide and candesartan changed from 8.9 to 80.59 hours, 11 to 69.20 hours and 9 to 101.49 hours respectively]. We have shown for the first time that encapsulating amlodipine, hydrochlorothiazide and candesartan into a single nanoformulation, to get the "NanoFDC (Fixed Dose Combination)" is a feasible strategy which aims to decrease pill burden.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H4759
Hydrochlorothiazide, crystalline
C7H8ClN3O4S2
H2910
Hydrochlorothiazide, meets USP testing specifications
C7H8ClN3O4S2