EMAIL THIS PAGE TO A FRIEND
International journal of cardiology
Continuous intravenous infusion of nicorandil for 4 hours before and 24 hours after percutaneous coronary intervention protects against contrast-induced nephropathy in patients with poor renal function.
PMID 26048382
Abstract
We conducted a prospective randomized trial to assess the protective effect of continuous intravenous infusion of nicorandil against contrast-induced nephropathy (CIN) in patients with poor renal function. We randomly assigned 213 patients who would subsequently undergo elective percutaneous coronary intervention (PCI) and who had a high serum cystatin C level to a saline group (n=107) or a nicorandil group (n=106, nicorandil infused in addition to saline for 4h before and 24h after PCI). There were no significant differences in baseline characteristics between the two groups. However, the average percent increases in serum creatinine and cystatin C following PCI were significantly smaller in the nicorandil group than the saline group. Likewise, the average percent decline in the estimated glomerular filtration rate was smaller in the nicorandil group. Correspondingly, the incidence of CIN was dramatically lower in the nicorandil group than the saline group (2.0% vs. 10.7%, p<0.02). Univariate regression analysis revealed nicorandil treatment to be the only significant predictor of CIN development (odds ratio: 0.173, 95% confidence interval: 0.037-0.812, p=0.026). Nicorandil strongly prevents CIN in patients with poor renal function undergoing PCI.