EMAIL THIS PAGE TO A FRIEND
Drug and alcohol dependence
Effects of chronic binge-like ethanol consumption on cocaine self-administration in rhesus monkeys.
PMID 26048636
Abstract
Most cocaine abusers also abuse alcohol, but little is known about interactions that promote co-abuse. These experiments in rhesus monkeys determined the effects of >8 weeks of ethanol (EtOH) consumption on cocaine self-administration (n=6), effects of dopamine (DA) receptor antagonists on cocaine reinforcement (n=3-4 per drug) and the ability of the D2-like DA receptor agonist quinpirole to elicit yawning (n=3). Monkeys self-administered cocaine (0.0-1.0mg/kg/injection, i.v.) under a 300-s fixed-interval schedule and the above-listed variables were measured before EtOH exposure. Next, monkeys consumed a sweetened, 4% EtOH solution in the home cage under binge-like conditions: 1h, 5 days/week with daily intake equaling 2.0g/kg EtOH. After approximately 8 weeks, measures were re-determined, then EtOH drinking was discontinued. Finally, acute effects of EtOH on cocaine self-administration were determined by infusing EtOH (0.0-1.0g/kg. i.v.) prior to cocaine self-administration sessions (n=4). In five of six monkeys, EtOH drinking increased self-administration of low cocaine doses but did not alter reinforcing effects of higher doses. Self-administration returned to baseline after EtOH access was terminated (n=3). Effects of DA receptor antagonists on cocaine self-administration were not consistently altered after EtOH consumption, but the ability of quinpirole to induce yawning was enhanced in two of three monkeys. Acute EtOH infusions only decreased self-administration of lower cocaine doses. Taken together, the data suggest that long-term EtOH exposure can increase sensitivity to cocaine, possibly by increasing D3 receptor sensitivity. Data do not support a role for acute pharmacological interactions in promoting cocaine/EtOH co-abuse.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
282227
Chlorhexidine, ≥99.5%
C22H30Cl2N10