EMAIL THIS PAGE TO A FRIEND
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine
Gabarapl1 mediates androgen-regulated autophagy in prostate cancer.
PMID 26050226
Abstract
Autophagy plays an important role in prostate cancer development. It promotes tumor cell survival and was found to be associated with androgen pathway. In the present study, we found that GABA(A) receptor-associated protein like 1 (Gabarapl1), a ubiquitin-like modifier, participates in the regulation of autophagy. Gabarapl1 is transcriptionally regulated by androgen receptor (AR) and has a repressive role in autophagy. Androgen deprivation downregulates Gabarapl1 in an AR dependent manner, resulting in the increase of autophagy flux. Elevated Gabarapl1 also represses the proliferation of prostate cancer cells. In summary, our study provides evidence to show that Gabarapl1 is a mediator involved in androgen-regulated autophagy process.