EMAIL THIS PAGE TO A FRIEND
Acta physiologica (Oxford, England)
Activation of angiotensin II type 1 receptors and contractile activity in human sigmoid colon in vitro.
PMID 26052867
Abstract
To analyse the effects of angiotensin II (Ang II) on the contractility of human sigmoid colon, and to characterize the subtype(s) of receptor(s) involved and the related action mechanism. The contractility of sigmoid colon circular muscle strips was recorded isometrically. RT-PCR and immunohistochemistry were used to reveal the eventual existence of a local renin-angiotensin system (RAS) and the distribution of Ang II receptors. Transcripts encoding for the Ang II type 1 (AT1 ) and the Ang II type 2 (AT2 ) receptor subtypes and for the angiotensin-converting enzyme in the whole-thickness muscular wall were observed. Ang II caused a concentration-dependent contractile response, which is antagonized by losartan, AT1 receptor antagonist, but not by PD123319, AT2 receptor antagonist. The joint application of losartan and PD123319 did not produce any additive effect. The contractile response to Ang II was partially reduced by tetrodotoxin, Na(+) voltage-gated neural channel blocker, and to some extent by SR48968, tachykinin NK2 receptor antagonist. However, hexamethonium, nicotinic receptor antagonist, atropine, cholinergic muscarinic receptor antagonist and SR140333, tachykinin NK1 receptor antagonist, were ineffective. Immunohistochemical analysis showed that AT1 receptors were expressed on the smooth muscle layers and myenteric plexus. Ang II positively modulates the spontaneous contractile activity of human sigmoid colon via activation of post-junctional and pre-junctional AT1 receptors, the latter located on the enteric nerves that modulate the release of tachykinins. The presence of the components of RAS in the human colon suggests that Ang II can be also locally generated to control colonic motility.
Related Materials