EMAIL THIS PAGE TO A FRIEND
Journal of Biomedical Materials Research Part A
Shape and surface chemistry effects on the cytotoxicity and cellular uptake of metallic nanorods and nanospheres.
PMID 26053238