EMAIL THIS PAGE TO A FRIEND
Environmental Toxicology and Chemistry / Setac
Fate of polychlorinated biphenyls in a contaminated lake ecosystem: combining equilibrium passive sampling of sediment and water with total concentration measurements of biota.
PMID 26053463