EMAIL THIS PAGE TO A FRIEND
Obesity (Silver Spring, Md.)
MFAP5 is related to obesity-associated adipose tissue and extracellular matrix remodeling and inflammation.
PMID 26054006
Abstract
Low-grade inflammation is involved in adipose tissue (AT) and extracellular matrix (ECM) remodeling and induces deposition of ECM proteins in AT. We have previously shown that MFAP5 (microfibrillar-associated protein 5) expression decreases in AT after weight loss. The aim of this study was to investigate MFAP5 localization in human AT and gene expression in adipocytes and the role of MFAP5 in adipocyte metabolism and inflammation. MFAP5 protein localization and gene expression were studied with immunohistochemistry and quantitative reverse transcriptase PCR (RT-qPCR) in human subcutaneous AT and cultured Simpson-Golabi-Behmel syndrome (SGBS) adipocytes, respectively. The effect of MFAP5 knock-down by siRNA on gene expression and insulin action was examined with RT-qPCR, western blot, and insulin-stimulated glucose uptake. The effect of different cytokines on MFAP5 gene and protein expression was investigated in cultured human SGBS preadipocytes. MFAP5 protein was highly expressed in AT, and gene expression decreased during adipocyte differentiation in SGBS cells. Treatment of preadipocytes with TNFα and TGFβ1 increased MFAP5 gene and protein expression. Furthermore, MFAP5 knock-down decreased the expression of genes involved in inflammation. Our results demonstrate that factors involving low-grade inflammation modulate MFAP5 expression and that the modified expression of MFAP5 may further regulate AT inflammation.