EMAIL THIS PAGE TO A FRIEND
BMC research notes
PCSK5 mutation in a patient with the VACTERL association.
PMID 26055999
Abstract
The VACTERL association is a typically sporadic, non-random collection of congenital anomalies that includes vertebral defects, anal atresia, cardiac defects, tracheoesophageal fistula with esophageal atresia, renal anomalies, and limb abnormalities. Although several chromosomal aberrations and gene mutations have been reported as disease-causative, these findings have been sparsely replicated to date. In the present study, whole exome sequencing of a case with the VACTERL association uncovered a novel frameshift mutation in the PCSK5 gene, which has been reported as one of the causative genes for the VACTERL association. Although this mutation appears potentially pathogenic in its functional aspects, it was also carried by the healthy father. Furthermore, a database survey revealed several other deleterious variants in the PCSK5 gene in the general population. Further studies are necessary to clarify the etiological role of the PCSK5 mutation in the VACTERL association.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A9251
Adenosine, ≥99%
C10H13N5O4
A4036
Adenosine, BioReagent, suitable for cell culture
C10H13N5O4
01890
Adenosine
C10H13N5O4